Learn More
TARGETMOL CHEMICALS INC CAY10595 5MG
SDP
Catalog No. 502581690
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. CAY10595 is an antagonist of the CRTh2 (DP2) receptor with a Ki of 10 nM. purity: 99%
Catalog No. 50-258-1690 Supplier TARGETMOL CHEMICALS INC Supplier No. T106915MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. CAY10595 is an antagonist of the CRTh2 (DP2) receptor with a Ki of 10 nM. purity: 99%