Learn More
Cayman Chemical CAY10602, Purity: ≥95%, CAS: 374922-43-7, 5 mg
SDP
Catalog No. 502888234
Encompass_Preferred
A SIRT1 activator; inhibits LPS-induced TNF-α release in THP-1 cells at 20 and 60 µM; decreases the expression of GPX4, SLC7A11, and SLC3A2, as well as increases the levels of iron and MDA, in A549, MDA-MB-231, and Hs 578T cells at 5 µM; reduces oleic acid-induced apoptosis and lipid accumulation in HepG2 cells when used at a concentration of 20 µM
Catalog No. 50-288-8234 Supplier Cayman Chemical Supplier No. 100097965 MG
A SIRT1 activator; inhibits LPS-induced TNF-α release in THP-1 cells at 20 and 60 µM; decreases the expression of GPX4, SLC7A11, and SLC3A2, as well as increases the levels of iron and MDA, in A549, MDA-MB-231, and Hs 578T cells at 5 µM; reduces oleic acid-induced apoptosis and lipid accumulation in HepG2 cells when used at a concentration of 20 µM