Learn More
Abcam CAY10602, SIRT1 Activator, 50MG
Catalog No. 502705767
Encompass_Preferred
MW 418.4 Da, Purity >95%. SIRT1 Activator. Suppresses NF-κB-dependent induction of TNF-α by LPS (IC₇₀ = 60 μM, THP-1 cells). Upregulates PPARδ-mediated transcription in ECs.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5767 Supplier Abcam Supplier No. AB14441650MG
MW 418.4 Da, Purity >95%. SIRT1 Activator. Suppresses NF-κB-dependent induction of TNF-α by LPS (IC₇₀ = 60 μM, THP-1 cells). Upregulates PPARδ-mediated transcription in ECs.
The product is subject to the following: Abcam Restricted Use Statement