Learn More
Medchemexpress LLC HY-18613 5mg Medchemexpress, CAY10603 CAS:1045792-66-2 Purity:>98%
SDP
Catalog No. 502030822
Encompass_Preferred
Medchemexpress, HY-18613 5mg CAY10603 CAS:1045792-66-2 CAY10603 (BML-281) is a potent and selective HDAC6 inhibitor, with an IC50 of 2 pM; CAY10603 (BML-281) also inhibits HDAC1, HDAC2, HDAC3, HDAC8, HDAC10, with IC50s of 271, 252, 0.42, 6851, 90.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0822 Supplier Medchemexpress LLC Supplier No. HY186135MG
Medchemexpress, HY-18613 5mg CAY10603 CAS:1045792-66-2 CAY10603 (BML-281) is a potent and selective HDAC6 inhibitor, with an IC50 of 2 pM; CAY10603 (BML-281) also inhibits HDAC1, HDAC2, HDAC3, HDAC8, HDAC10, with IC50s of 271, 252, 0.42, 6851, 90.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.