Learn More
STEMCELL Technologies CAY10603, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 73582
Encompass
CAY10603 is a selective and potent inhibitor of histone deacetylase 6 (HDAC6; IC₅₀ = 0.002 nM; Kozikowski et al.). HDAC6 deacetylates lysine residues on the N-terminal part of histones. HDAC6 is also known to regulate heat shock protein 90 (hsp90) via hyperacetylation (Rao et al.).CANCER RESEARCH· Inhibits growth of several pancreatic cancer cell lines (IC₅₀ = 0.1 - 1 μM) and SCCOHT, an ovarian cancer cell line (Kozikowski et al.; Wang et al.).
Catalog No. 50-197-6099