Learn More
Abcam CAY10626, PI3Kalpha/mTOR inhibitor, 5MG
Catalog No. 502707145
Encompass_Preferred
MW 624.66 g/mol, Purity >98%. Potent, dual PI3Kα/mTOR inhibitor (IC₅₀ values are 0.9 and 0.6 nM at PI3Kα and mTOR, respectively). Inhibits tumor cell growth, suppresses phosphorylation of downstream targets of PI3Kα and mTOR in vitro and promotes tumor regression in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7145 Supplier Abcam Supplier No. AB1209035MG
MW 624.66 g/mol, Purity >98%. Potent, dual PI3Kα/mTOR inhibitor (IC₅₀ values are 0.9 and 0.6 nM at PI3Kα and mTOR, respectively). Inhibits tumor cell growth, suppresses phosphorylation of downstream targets of PI3Kα and mTOR in vitro and promotes tumor regression in vivo..
The product is subject to the following: Abcam Restricted Use Statement