Learn More
TARGETMOL CHEMICALS INC CAY10650 5MG
SDP
Catalog No. 502578828
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CAY10650 is an effective inhibitor of cytosolic phospholipase A2(alpha) (cPLA2(alpha) IC50 12 nM). purity: 95%
Catalog No. 50-257-8828 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ01055MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CAY10650 is an effective inhibitor of cytosolic phospholipase A2(alpha) (cPLA2(alpha) IC50 12 nM). purity: 95%