Learn More
Cayman Chemical CAY10729 trifluoroacetate sa
SDP
Catalog No. 502935439
Encompass_Preferred
A tetrazole derivative of Cl-amidine; selectively inhibits PAD1 over PAD2, PAD3, and PAD4 (kinact/KI = 162,200, 60,000, 17,500, and 25,400 M-1min-1 for PAD1-4, respectively)
Catalog No. 50-293-5439 Supplier Cayman Chemical Supplier No. 26546500 UG
A tetrazole derivative of Cl-amidine; selectively inhibits PAD1 over PAD2, PAD3, and PAD4 (kinact/KI = 162,200, 60,000, 17,500, and 25,400 M-1min-1 for PAD1-4, respectively)