Learn More
Cayman Chemical CAY10740hydrochlorIde 10mg
SDP
Catalog No. 502937746
Encompass_Preferred
An analog of GSK484; decreases the viability of 4T1 cells by 85 and 98% at 5 and 10 μM, respectively, which is more than five-fold as effective as GSK484
Catalog No. 50-293-7746 Supplier Cayman Chemical Supplier No. 2832010 MG
An analog of GSK484; decreases the viability of 4T1 cells by 85 and 98% at 5 and 10 μM, respectively, which is more than five-fold as effective as GSK484