Learn More
Cayman Chemical Cayman Chemical BSA-Oleate Reagent Set, 5 mM, Cell Culture
SDP
Catalog No. NC3983043
Encompass_Preferred
The BSA-Oleate Reagent Set provides a 5 mM solution of BSA-oleate complex and a fatty acid-free BSA control. This set is designed for efficient fatty acid delivery to cells in culture, suitable for monitoring fatty acid oxidation and other metabolic studies.
  • Contains a 5 mM BSA-oleate complex and a fatty acid-free BSA control.
  • BSA-oleate complex is composed of oleic acid and fatty acid-free bovine serum albumin (BSA) at an an approximate 6:1 molar ratio.
  • Prepared under sterile conditions and filtered into sterile vials.
  • Supports short- and long-term cell culture applications (25+ hours).
Catalog No. NC3983043 Supplier Cayman Chemical Supplier No. 44712
The BSA-Oleate Reagent Set provides a 5 mM solution of BSA-oleate complex and a fatty acid-free BSA control. This set is designed for efficient fatty acid delivery to cells in culture, suitable for monitoring fatty acid oxidation and other metabolic studies.
  • Contains a 5 mM BSA-oleate complex and a fatty acid-free BSA control.
  • BSA-oleate complex is composed of oleic acid and fatty acid-free bovine serum albumin (BSA) at an an approximate 6:1 molar ratio.
  • Prepared under sterile conditions and filtered into sterile vials.
  • Supports short- and long-term cell culture applications (25+ hours).