Learn More
Cayman Chemical Cayman Practice ELISA Kit
SDP
Supplier:  Cayman Chemical 1000965896 WELL
Encompass_Preferred
This assay has been developed for researchers that do not have experience performing ELISAs. It can be used as a practice tool allowing you to become comfortable with running Cayman ELISAs. This kit contains enough reagents to run at least four complete standard curves.
Catalog No. 50-288-7075