Learn More
Cayman Chemical Caymn LIpIDOT StrIps- Deve 1ea
SDP
Catalog No. 502918637
Encompass_Preferred
Cayman LipiDOT Strips™ - Developing Chamber can be used with Cayman LipiDOT Strips™ - PIPs Plus (Item No. 38924) for the blocking, incubation, and washing steps. Box Dimensions: 2 7/8"W x 2"L x 1 1/4"H
Catalog No. 50-291-8637 Supplier Cayman Chemical Supplier No. 393801 EA
Cayman LipiDOT Strips™ - Developing Chamber can be used with Cayman LipiDOT Strips™ - PIPs Plus (Item No. 38924) for the blocking, incubation, and washing steps. Box Dimensions: 2 7/8"W x 2"L x 1 1/4"H