Learn More
Cayman Chemical Caymn LIpIDOT StrIps- PIPs 2ea
SDP
Catalog No. 502915663
Encompass_Preferred
A simple tool for rapid identification of protein-lipid interactions; contains 22 biologically relevant lipids and two blank samples for use as controls; lipids and controls are spotted on a 3.175 cm x 6 cm supported nitrocellulose membrane in a 3 x 8 grid; intended for use with Cayman LipiDOT Strips™ Developing Chamber
Catalog No. 50-291-5663 Supplier Cayman Chemical Supplier No. 389242 EA
A simple tool for rapid identification of protein-lipid interactions; contains 22 biologically relevant lipids and two blank samples for use as controls; lipids and controls are spotted on a 3.175 cm x 6 cm supported nitrocellulose membrane in a 3 x 8 grid; intended for use with Cayman LipiDOT Strips™ Developing Chamber