Learn More
Braintree Scientific Gemini Cautery System, Dual Operating Temps
SDP
Catalog No. NC9183895
Encompass
Gemini Cautery System - Dual Operating Temperatures. Battery operated system provides two different operating temperatures: 1300F and 2200F. Easily replaceable, autoclavable, nickel-chrome tips require less power and provide longer battery life. Convenient battery spacer converts hand piece from high temp to low temp. Comes complete with standard AA batteries, battery spacer, 2 each tips - sizes 5mm and 10mm, all in a hard plastic case with die cut foam for convenient storage. **Sold for research purposes only and not for human use**
Catalog No. NC9183895 Supplier Braintree Scientific Supplier No. GEM5917
Gemini Cautery System - Dual Operating Temperatures. Battery operated system provides two different operating temperatures: 1300F and 2200F. Easily replaceable, autoclavable, nickel-chrome tips require less power and provide longer battery life. Convenient battery spacer converts hand piece from high temp to low temp. Comes complete with standard AA batteries, battery spacer, 2 each tips - sizes 5mm and 10mm, all in a hard plastic case with die cut foam for convenient storage. **Sold for research purposes only and not for human use**