Learn More
Medchemexpress LLC HY-135145 50mg , CB-103 CAS:218457-67-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13514550MG
Encompass_Preferred
HY-135145 50mg CB-103 CAS: 218457-67-1 CB-103 is a first-in-class, orally active protein-protein interaction (PPI) inhibitor of the NOTCH transcriptional activation complex. CB-103 has anti-tumor activity[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0765