Learn More
As One International Inc 10x Combination Buffer, 10 x 1.5 ml
Catalog No. 501655976
Encompass_Preferred
Combination Buffer offers a balanced ammonium-potassium formulation that result in tolerance toward optimization of magnesium concentration and primer annealing temperatures.
Catalog No. 50-165-5976 Supplier As One International Inc Supplier No. AO303110
Combination Buffer offers a balanced ammonium-potassium formulation that result in tolerance toward optimization of magnesium concentration and primer annealing temperatures.