Learn More
Apexbio Technology LLC CB-5083, 1542705-92-9, MFCD28963914, 10 mg
SDP
Supplier:  Apexbio Technology LLC B603210
Encompass_Preferred
MF: C24H23N5O2, MW: 413.47, Solubility: 20.65mg/mL in DMSO. CB-5083 is an orally bioavailable inhibitor of p97. P97 is an AAA-ATPase involved in multiple cellular functions such as organelle membrane homotypic fusion and sorting of endosomal cargo. P97 is also known as valosin-containing protein which plays important roles in regulating protein homeostasis.
Catalog No. 50-115-2550