Learn More
Selleck Chemical LLC CB-5083 5mg 1542705-92-9
SDP
Catalog No. 501365653
Encompass_Preferred
CB-5083 is a potent, selective, and orally bioavailable p97 AAA ATPase inhibitor with IC50 of 11 nM. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5653 Supplier Selleck Chemical LLC Supplier No. S81015MG
CB-5083 is a potent, selective, and orally bioavailable p97 AAA ATPase inhibitor with IC50 of 11 nM. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.