Learn More
Medchemexpress LLC CB-6644 5mg | 2316817-88-4 | 5 MG
SDP
Catalog No. 5000308077
Encompass_Preferred
CB-6644 is a selective allosteric inhibitor of the RUVBL1/2 ATPase with a reported IC50 of 15 nM. It exhibits antiproliferative activity in multiple cancer cell lines and oral antitumor efficacy in xenograft models.
  • Selective inhibitor of the RUVBL1/2 complex
  • Blocks ATPase activity with an IC50 of 15 nM
  • Demonstrates antiproliferative activity in vitro
  • Shows oral antitumor efficacy in xenograft models
  • Solid, white to light yellow appearance
  • Soluble in DMSO (100 mg/mL); in vivo formulations reported at ≥ 2.08 mg/mL
  • Store solid at -20°C under nitrogen; solutions at -80°C for long-term storage
Catalog No. 50-003-08077 Supplier Medchemexpress LLC Supplier No. HY1144295MG
CB-6644 is a selective allosteric inhibitor of the RUVBL1/2 ATPase with a reported IC50 of 15 nM. It exhibits antiproliferative activity in multiple cancer cell lines and oral antitumor efficacy in xenograft models.
  • Selective inhibitor of the RUVBL1/2 complex
  • Blocks ATPase activity with an IC50 of 15 nM
  • Demonstrates antiproliferative activity in vitro
  • Shows oral antitumor efficacy in xenograft models
  • Solid, white to light yellow appearance
  • Soluble in DMSO (100 mg/mL); in vivo formulations reported at ≥ 2.08 mg/mL
  • Store solid at -20°C under nitrogen; solutions at -80°C for long-term storage