Learn More
Medchemexpress LLC HY-12248 100mg Medchemexpress, CB-839 CAS:1439399-58-2 Purity:98%
SDP
Catalog No. 501637696
Encompass_Preferred
Medchemexpress,HY-12248 100mg CB-839 CAS:1439399-58-2 Formula:C26H24F3N7O3S CB-839 is a potent and selective inhibitor of glutaminase with an IC50 of less than 50 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7696 Supplier Medchemexpress LLC Supplier No. HY12248100MG
Medchemexpress,HY-12248 100mg CB-839 CAS:1439399-58-2 Formula:C26H24F3N7O3S CB-839 is a potent and selective inhibitor of glutaminase with an IC50 of less than 50 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.