Learn More
Selleck Chemical LLC Telaglenastat (CB-839) 5mg 1439399-58-2
SDP
Supplier:  Selleck Chemical LLC S76555MG
Encompass_Preferred
Telaglenastat (CB-839) is a potent, selective, and orally bioavailable glutaminase inhibitor with IC50 of 24 nM for recombinant human GAC. CB-839(Telaglenastat) inudces autophagy and has antitumor activity. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5083