Learn More
Adam Equipment CB 3000 Compact Balance 3000g capacity x 1g readability
Supplier:  Adam Equipment CB 3000
Encompass_Preferred
Adam Equipment CB Compact Balances can manage most small tasks, such as measuring compounds in chemistry classes, portioning at food service establishments, or weighing small animals. Compact and portable, the CB can easily be transported to the field, or from bench to bench in a science classroom. The CB is powered by battery or an optional AC adapter. A sealed keypad and one-piece housing make the CB simple to clean. The grade 304 stainless steel pan can be wiped down quickly and accommodates either single items or bulk powders. The CB compact scale boasts best-in-class performance and value. Its grade 304 stainless steel pan, sealed keypad and one-piece housing are easy to clean, while built-in battery operation means it can be used anywhere. Operates with AC adapter (not included) or batteries (included).
Catalog No. NC2611861