Learn More
Charles River Labs Cell Solutions Cord Blood CD34+ Stem/Progenitor Cells, 1 Million cells, Cryopreserved
Catalog No. 502256142
Encompass
CD34+ hematopoietic stem cells are positively selected from freshly collected cord blood. After isolation, CD34+ stem cells are evaluated for purity and viability by flow cytometry.
This item is not returnable. View return policy
Catalog No. 50-225-6142 Supplier Charles River Labs Cell Solutions Supplier No. CB34C3
Add to Cart
Add to Cart
CD34+ hematopoietic stem cells are positively selected from freshly collected cord blood. After isolation, CD34+ stem cells are evaluated for purity and viability by flow cytometry.