Learn More
As One International Inc 10x Combination Buffer, Magnesium free and Tween free, 10 x 1.5 ml
Catalog No. 501655979
Encompass_Preferred
Combination Buffer offers a balanced ammonium-potassium formulation that result in tolerance toward optimization of magnesium concentration and primer annealing temperatures. Detergent (Tween or Triton) free buffers are ideal for automation and downstream applications using fluorescent spectrometry. Magnesium free buffers are recommended if you want to optimize the magnesium concentration.
Catalog No. 50-165-5979 Supplier As One International Inc Supplier No. AO303510
Combination Buffer offers a balanced ammonium-potassium formulation that result in tolerance toward optimization of magnesium concentration and primer annealing temperatures. Detergent (Tween or Triton) free buffers are ideal for automation and downstream applications using fluorescent spectrometry. Magnesium free buffers are recommended if you want to optimize the magnesium concentration.