Learn More
As One International Inc 10x Combination Buffer, Tween free, 10 x 1.5 ml
Catalog No. 501655978
Encompass_Preferred
Combination Buffer offers a balanced ammonium-potassium formulation that result in tolerance toward optimization of magnesium concentration and primer annealing temperatures. Detergent (Tween or Triton) free buffers are ideal for automation and downstream applications using fluorescent spectrometry.
Catalog No. 50-165-5978 Supplier As One International Inc Supplier No. AO303410
Combination Buffer offers a balanced ammonium-potassium formulation that result in tolerance toward optimization of magnesium concentration and primer annealing temperatures. Detergent (Tween or Triton) free buffers are ideal for automation and downstream applications using fluorescent spectrometry.