Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CB096 | 108883-90-5 | 98.7% | 285.25 g·mol⁻¹ | C14H11N3O4 | 250 MG
SDP

Catalog No. 5000282807
Encompass_Preferred

CB096 is a small-molecule RNA translation inhibitor that binds the 5′CGG/3′GGC internal loop of r(G4C2)exp RNA, disrupting its gain-of-function mechanism and reducing production of toxic dipeptide repeats. It inhibits RAN translation in cell assays (IC50 = 20 μM in HEK293T) and modulates RNA splicing and nucleocytoplasmic transport.

  • Inhibits RAN translation in cells (IC50 = 20 μM in HEK293T).
  • Binds the 5′CGG/3′GGC internal loop of r(G4C2)exp RNA.
  • High purity of 98.72%.
  • Solid, yellow to orange appearance.
  • Soluble in DMSO at 50 mg/mL.
  • Storage: sealed, away from moisture; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple quantities for research use.

Catalog No. 50-002-82807 Supplier Medchemexpress LLC Supplier No. HYW141374250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CB096 is a small-molecule RNA translation inhibitor that binds the 5′CGG/3′GGC internal loop of r(G4C2)exp RNA, disrupting its gain-of-function mechanism and reducing production of toxic dipeptide repeats. It inhibits RAN translation in cell assays (IC50 = 20 μM in HEK293T) and modulates RNA splicing and nucleocytoplasmic transport.

  • Inhibits RAN translation in cells (IC50 = 20 μM in HEK293T).
  • Binds the 5′CGG/3′GGC internal loop of r(G4C2)exp RNA.
  • High purity of 98.72%.
  • Solid, yellow to orange appearance.
  • Soluble in DMSO at 50 mg/mL.
  • Storage: sealed, away from moisture; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.