Learn More
Medchemexpress LLC HY-116649 5mg Medchemexpress, CB1 antagonist 2 CAS:614726-85-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1166495MG
Encompass_Preferred
Medchemexpress, HY-116649 5mg CB1 antagonist 2 CAS:614726-85-1 CB1 antagonist 2 is caimabinoid 1 (CB1) antagonist extracted from patent WO2016184310A1, compound 3, inhibits CB1 in vivo with an IC50 of 25.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9435