Learn More
Medchemexpress LLC CB1 inverse agonist 2 | 1019839-52-1 | 99.2% | 5 MG
SDP
Catalog No. 5000233990
Encompass_Preferred
CB1 inverse agonist 2 is an orally active inverse agonist of the cannabinoid CB1 receptor shown to inhibit CP55940-induced hypothermia and anorexia in mouse models. Supplied at high purity for research applications, it is appropriate for preclinical studies and small-scale pharmacology work.
  • Orally active CB1 receptor inverse agonist.
  • Demonstrated efficacy against CP55940-induced hypothermia and anorexia in mice.
  • High purity suitable for research use.
  • Molecular formula C24H20ClFN2OS and molecular weight 438.94 g/mol.
  • Available in small quantities for preclinical and exploratory studies.
Catalog No. 50-002-33990 Supplier Medchemexpress LLC Supplier No. HY120955MG
CB1 inverse agonist 2 is an orally active inverse agonist of the cannabinoid CB1 receptor shown to inhibit CP55940-induced hypothermia and anorexia in mouse models. Supplied at high purity for research applications, it is appropriate for preclinical studies and small-scale pharmacology work.
  • Orally active CB1 receptor inverse agonist.
  • Demonstrated efficacy against CP55940-induced hypothermia and anorexia in mice.
  • High purity suitable for research use.
  • Molecular formula C24H20ClFN2OS and molecular weight 438.94 g/mol.
  • Available in small quantities for preclinical and exploratory studies.