Learn More
Enzo Life Sciences Cannabinoid receptor 1 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210316R100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant rat CB1 (cannabinoid receptor 1) (aa 1-77). UniProt ID: P20272. Species reactivity: Human, Mouse, Rat, Monkey, Fish, Chicken, Amphibian. Applications: Flow Cytometry, ICC, IHC (PS), WB. Recommended Dilutions: Immunocytochemistry (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Affinity purified IgG. Formulation: Liquid. In PBS containing 50% glycerol, 1mg/ml BSA and 0.05% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8110