Learn More
Cayman Chemical CB1 Receptor (C-Term) Blocking Peptide, 1 ea
SDP
Catalog No. 502886888
Encompass_Preferred
Peptide Sequence: CB1 receptor (C-Term) amino acids 461-472 · To be used in conjunction with Cayman’s CB1 Receptor (C-Term) polyclonal antibody (Item No. 10006590) to block protein-antibody complex formation during immunochemical analysis of CB1
Catalog No. 50-288-6888 Supplier Cayman Chemical Supplier No. 100065911 EA
Peptide Sequence: CB1 receptor (C-Term) amino acids 461-472 · To be used in conjunction with Cayman’s CB1 Receptor (C-Term) polyclonal antibody (Item No. 10006590) to block protein-antibody complex formation during immunochemical analysis of CB1