Learn More
Cayman Chemical CB1 RECPTR Blockng PeptID
SDP
Catalog No. 502978865
Encompass_Preferred
To be used in conjunction with Cayman’s CB1 receptor polyclonal antibody (Item No. 101500) to block protein-antibody complex formation during immunochemical analysis for the CB1 receptor.
Catalog No. 50-297-8865 Supplier Cayman Chemical Supplier No. 301500200 UG
To be used in conjunction with Cayman’s CB1 receptor polyclonal antibody (Item No. 101500) to block protein-antibody complex formation during immunochemical analysis for the CB1 receptor.