Learn More
Labjack Corporation CB15 Terminal Board
SDP
Catalog No. NC3357522
Encompass
The CB15 provides convenient screw terminals for the 12 digital I/O lines on the DB15 connector of compatible LabJack devices. Includes a screwdriver. Not compatible with the U12.
Catalog No. NC3357522 Supplier Labjack Corporation Supplier No. CB15
The CB15 provides convenient screw terminals for the 12 digital I/O lines on the DB15 connector of compatible LabJack devices. Includes a screwdriver. Not compatible with the U12.