Learn More
Selleck Chemical LLC CB1954 10mg 21919-05-1 Tretazicar
SDP
Catalog No. 501365310
Encompass_Preferred
CB1954(Tretazicar) is a anticancer prodrug that is converted in the presence of the enzyme NQO2 and co-substrate caricotamide ( EP-0152R) (EP) into a potent cytotoxic bifunctional alkylating agent. It can be activated by NAD(P)H quinone oxidoreductase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5310 Supplier Selleck Chemical LLC Supplier No. S782910MG
CB1954(Tretazicar) is a anticancer prodrug that is converted in the presence of the enzyme NQO2 and co-substrate caricotamide ( EP-0152R) (EP) into a potent cytotoxic bifunctional alkylating agent. It can be activated by NAD(P)H quinone oxidoreductase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.