Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CB2 modulator 1 | 666261-80-9 | 99.8% | 380.36 | C18H19F3N4O2 | 5 MG
SDP

Catalog No. 5000223346
Encompass_Preferred

CB2 modulator 1 is a potent small-molecule modulator of the cannabinoid receptor CB2 intended for research use. It has reported activity relevant to immune disorders, inflammation, osteoporosis, and renal ischemia. Supplied as a high-purity solid for in vitro and exploratory studies; follow appropriate safety and regulatory guidance.

  • Acts on cannabinoid receptor CB2.
  • Reported activity in immune disorders, inflammation, osteoporosis, and renal ischemia.
  • CAS number 666261-80-9.
  • Molecular formula C18H19F3N4O2.
  • Molecular weight 380.36 g/mol.
  • Purity 99.75% (reported).
  • Available in small pack sizes for screening, e.g., 5 mg.
  • Intended for research use only; follow appropriate safety guidance.

Catalog No. 50-002-23346 Supplier Medchemexpress LLC Supplier No. HY1354195MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CB2 modulator 1 is a potent small-molecule modulator of the cannabinoid receptor CB2 intended for research use. It has reported activity relevant to immune disorders, inflammation, osteoporosis, and renal ischemia. Supplied as a high-purity solid for in vitro and exploratory studies; follow appropriate safety and regulatory guidance.

  • Acts on cannabinoid receptor CB2.
  • Reported activity in immune disorders, inflammation, osteoporosis, and renal ischemia.
  • CAS number 666261-80-9.
  • Molecular formula C18H19F3N4O2.
  • Molecular weight 380.36 g/mol.
  • Purity 99.75% (reported).
  • Available in small pack sizes for screening, e.g., 5 mg.
  • Intended for research use only; follow appropriate safety guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.