Learn More
Enzo Life Sciences Cannabinoid receptor 2 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210315R100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human CB2 (cannabinoid receptor 2) (aa 1-33). Species reactivity: Human, Rat. Applications: Flow Cytometry, ICC, IHC (FS), IHC (PS), WB. Purity: Epitope-affinity purified IgG. Formulation: Liquid. In PBS containing 50% glycerol, 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-200-8109