Learn More
TARGETMOL CHEMICALS INC CB2 RECEPTOR AGONIST 2 5MG
SDP
Catalog No. 502580987
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. CB2 receptor agonist 2 (ZINC72105556) is a potent and selective agonist for the CB2 (cannabinoid type 2) receptor Ki = 8.5 nM. CB2 receptor agonist 2 has high affinity and selectivity for CB2. purity: 98%
Catalog No. 50-258-0987 Supplier TARGETMOL CHEMICALS INC Supplier No. T220095MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. CB2 receptor agonist 2 (ZINC72105556) is a potent and selective agonist for the CB2 (cannabinoid type 2) receptor Ki = 8.5 nM. CB2 receptor agonist 2 has high affinity and selectivity for CB2. purity: 98%