Learn More
Cayman Chemical CB2 RECPTR Blockng PeptID
SDP
Catalog No. 502978864
Encompass_Preferred
Peptide Sequence: human CB2 receptor sequence amino acids 20-33 (NPMKDYMILSGPQK) · To be used in conjunction with Cayman’s CB2 Receptor Polyclonal Antibody (Catalog No. 101550) to block protein-antibody complex formation during immunochemical analysis for the CB2 receptor. · The CB2 receptor is localized predominantly in peripheral tissues, including the spleen and hemopoietic cells.
Catalog No. 50-297-8864 Supplier Cayman Chemical Supplier No. 301550200 UG
Peptide Sequence: human CB2 receptor sequence amino acids 20-33 (NPMKDYMILSGPQK) · To be used in conjunction with Cayman’s CB2 Receptor Polyclonal Antibody (Catalog No. 101550) to block protein-antibody complex formation during immunochemical analysis for the CB2 receptor. · The CB2 receptor is localized predominantly in peripheral tissues, including the spleen and hemopoietic cells.