Learn More
Eurofins Discoverx Ready-to-Assay CB2 Cannabinoid Receptor Frozen Cells, 2 Vials of Cells. (HTS020RTA)
Catalog No. 502005682
Encompass
The Ready-to-Assay CB2 Cannabinoid Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. adrenocorticotropin) based activation of CNR2 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CNR2 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of CNR2, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5682 Supplier Eurofins Discoverx Supplier No. HTS020RTA
The Ready-to-Assay CB2 Cannabinoid Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. adrenocorticotropin) based activation of CNR2 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CNR2 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of CNR2, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.