Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cb2r/faah modulator-2 | 2876918-68-0 | 367.52 g·mol⁻¹ | C24H33NO2 | 10 MG
SDP

Catalog No. 5000285902
Encompass_Preferred

CB2R/FAAH modulator-2 is a research-grade small-molecule dual-target modulator that acts as an agonist at the cannabinoid CB2 receptor and an inhibitor of fatty acid amide hydrolase (FAAH). It is supplied as a solid for in vitro and in vivo pharmacology studies involving cannabinoid signaling and endocannabinoid metabolism. The compound has CAS 2876918-68-0, molecular formula C24H33NO2, and molecular weight 367.52 g·mol⁻¹.

  • Dual CB2R agonist and FAAH inhibitor.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Provided as a solid, with an option as 10 mM in DMSO solution.
  • Available in small research sizes, for example 10 mg.
  • CAS number 2876918-68-0 for unambiguous identification.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.

Catalog No. 50-002-85902 Supplier Medchemexpress LLC Supplier No. HY15225310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CB2R/FAAH modulator-2 is a research-grade small-molecule dual-target modulator that acts as an agonist at the cannabinoid CB2 receptor and an inhibitor of fatty acid amide hydrolase (FAAH). It is supplied as a solid for in vitro and in vivo pharmacology studies involving cannabinoid signaling and endocannabinoid metabolism. The compound has CAS 2876918-68-0, molecular formula C24H33NO2, and molecular weight 367.52 g·mol⁻¹.

  • Dual CB2R agonist and FAAH inhibitor.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Provided as a solid, with an option as 10 mM in DMSO solution.
  • Available in small research sizes, for example 10 mg.
  • CAS number 2876918-68-0 for unambiguous identification.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.