Learn More
Medchemexpress LLC CB2R/FAAH modulator-2 | 2876918-68-0 | C24H33NO2 | 1 ML
SDP
Catalog No. 5000372997
Encompass_Preferred
CB2R/FAAH modulator-2 is a dual targeting modulator functioning as a CB2R agonist and FAAH inhibitor. It has Ki values of 10.8 nM for CB2R and 152.9 nM for CB1R, and an IC50 value of 6.2 μM for FAAH. This modulator can be utilized in studies related to cancer, neurodegenerative diseases, and COVID-19 infection. In vitro, it has been shown to reduce the production of pro-inflammatory cytokines such as TNFα, IFN-γ, IL-1 ß, and IL6 in unstimulated monocytes and macrophages.
  • Acts as a CB2R agonist
  • Functions as a FAAH inhibitor
  • Relevant for studies on cancer
  • Useful in research on neurodegenerative diseases
  • Applicable to COVID-19 infection studies
  • Reduces production of pro-inflammatory cytokines
Catalog No. 50-003-72997 Supplier Medchemexpress LLC Supplier No. HY15225310MM/1ML
CB2R/FAAH modulator-2 is a dual targeting modulator functioning as a CB2R agonist and FAAH inhibitor. It has Ki values of 10.8 nM for CB2R and 152.9 nM for CB1R, and an IC50 value of 6.2 μM for FAAH. This modulator can be utilized in studies related to cancer, neurodegenerative diseases, and COVID-19 infection. In vitro, it has been shown to reduce the production of pro-inflammatory cytokines such as TNFα, IFN-γ, IL-1 ß, and IL6 in unstimulated monocytes and macrophages.
  • Acts as a CB2R agonist
  • Functions as a FAAH inhibitor
  • Relevant for studies on cancer
  • Useful in research on neurodegenerative diseases
  • Applicable to COVID-19 infection studies
  • Reduces production of pro-inflammatory cytokines