Learn More
Medchemexpress LLC CB2R/FAAH modulator-3 | 2876918-67-9 | C22H31NO2 | 25 MG
SDP
Catalog No. 5000375367
Encompass_Preferred
CB2R/FAAH modulator-3 is a dual-targeting modulator that acts as a CB2R agonist and FAAH inhibitor. It has potential applications in studies related to cancer, neurodegenerative diseases, and COVID-19 infection.
  • Acts as a dual-targeting modulator
  • Functions as a CB2R agonist and FAAH inhibitor
  • Exhibits Ki values of 20.1 nM for CB2R and 67.6 nM for CB1R
  • Has an IC50 value of 3.4 μM for FAAH
  • Reduces pro-inflammatory cytokines such as TNFα, IFN-γ, IL-1β, and IL6 in unstimulated monocytes and macrophages at 10 μM
Catalog No. 50-003-75367 Supplier Medchemexpress LLC Supplier No. 25MGHY152254
CB2R/FAAH modulator-3 is a dual-targeting modulator that acts as a CB2R agonist and FAAH inhibitor. It has potential applications in studies related to cancer, neurodegenerative diseases, and COVID-19 infection.
  • Acts as a dual-targeting modulator
  • Functions as a CB2R agonist and FAAH inhibitor
  • Exhibits Ki values of 20.1 nM for CB2R and 67.6 nM for CB1R
  • Has an IC50 value of 3.4 μM for FAAH
  • Reduces pro-inflammatory cytokines such as TNFα, IFN-γ, IL-1β, and IL6 in unstimulated monocytes and macrophages at 10 μM