Learn More
Medchemexpress LLC CB2R/FAAH modulator-3 | 2876918-67-9 | C22H31NO2 | 5 MG
SDP
Catalog No. 5000373248
Encompass_Preferred
CB2R/FAAH modulator-3 (compound 27) is a dual-targeting modulator that acts as a CB2R agonist and FAAH inhibitor. It has Ki values of 20.1 nM for CB2R and 67.6 nM for CB1R, and an IC50 value of 3.4 μM for FAAH. This compound can be used in studies related to cancer, deleterious inflammatory cascades in neurodegenerative diseases, and COVID-19 infection.
  • Dual-targeting modulator: acts as a CB2R agonist and FAAH inhibitor.
  • Can be used in studies related to cancer, deleterious inflammatory cascades occurring in neurodegenerative diseases, and COVID-19 infection.
  • At 10 μM, it reduces the production of the pro-inflammatory cytokines TNFα, IFN-γ, IL-1β, and IL6 in unstimulated monocytes and macrophages.
Catalog No. 50-003-73248 Supplier Medchemexpress LLC Supplier No. HY1522545MG
CB2R/FAAH modulator-3 (compound 27) is a dual-targeting modulator that acts as a CB2R agonist and FAAH inhibitor. It has Ki values of 20.1 nM for CB2R and 67.6 nM for CB1R, and an IC50 value of 3.4 μM for FAAH. This compound can be used in studies related to cancer, deleterious inflammatory cascades in neurodegenerative diseases, and COVID-19 infection.
  • Dual-targeting modulator: acts as a CB2R agonist and FAAH inhibitor.
  • Can be used in studies related to cancer, deleterious inflammatory cascades occurring in neurodegenerative diseases, and COVID-19 infection.
  • At 10 μM, it reduces the production of the pro-inflammatory cytokines TNFα, IFN-γ, IL-1β, and IL6 in unstimulated monocytes and macrophages.