Learn More
TARGETMOL CHEMICALS INC CB2R/FAAH MODULATOR-3 5MG
SDP
Catalog No. 502579265
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CB2R/FAAH modulator-3 (compound 27) is a modulator targeting at CB2R and FAAH. CB2R/FAAH modulator-3 acts as a CB2R agonist and FAAH inhibitor the Ki values are 20.1 and 67.6 nM for CB2R and CB1R respectively and the IC50 value for FAAH is 3.4 uM. CB2R/FAAH modulator-3 has research value related to cancer deleterious inflammatory cascades occurring in neurodegenerative diseases and COVID-19 infection. purity: 99%
Catalog No. 50-257-9265 Supplier TARGETMOL CHEMICALS INC Supplier No. T677475MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CB2R/FAAH modulator-3 (compound 27) is a modulator targeting at CB2R and FAAH. CB2R/FAAH modulator-3 acts as a CB2R agonist and FAAH inhibitor the Ki values are 20.1 and 67.6 nM for CB2R and CB1R respectively and the IC50 value for FAAH is 3.4 uM. CB2R/FAAH modulator-3 has research value related to cancer deleterious inflammatory cascades occurring in neurodegenerative diseases and COVID-19 infection. purity: 99%