Learn More
TARGETMOL CHEMICALS INC CB2R/FAAHMODULATOR-2 5MG
Supplier: TARGETMOL CHEMICALS INC T677455MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CB2R/FAAH modulator-2 (compound 26) is a dual modulator targeting at CB2R and FAAH. CB2R/FAAH modulator-2 acts as a CB2R agonist and FAAH inhibitor the Ki values are 10.8 and 152.9 nM for CB2R and CB1R respectively and the IC50 value for FAAH is 6.2 uM. CB2R/FAAH modulator-2 has studies research value related to cancer deleterious inflammatory cascades occurring in neurodegenerative diseases and COVID-19 infection. purity: 99%