Learn More
Medchemexpress LLC HY-14373 5mg Medchemexpress, CB30865 CAS:206275-15-2 Purity:>98%
SDP
Catalog No. 502081594
Encompass_Preferred
Medchemexpress, HY-14373 5mg CB30865 CAS:206275-15-2 CB30865 (ZM 242421) is a nicotinamide phosphoribosyltransferase (Nampt) inhibitor, with potent cytotoxicity. CB30865 is highly potent against a variety of human tumour cell lines (IC50 values in the 1-10 nM range). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1594 Supplier Medchemexpress LLC Supplier No. HY143735MG
Medchemexpress, HY-14373 5mg CB30865 CAS:206275-15-2 CB30865 (ZM 242421) is a nicotinamide phosphoribosyltransferase (Nampt) inhibitor, with potent cytotoxicity. CB30865 is highly potent against a variety of human tumour cell lines (IC50 values in the 1-10 nM range). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.