Learn More
Labjack Corporation CB37 Terminal Board
SDP
Catalog No. NC3357520
Encompass
Provides convenient screw terminals for the DB37 connector on the LabJack U6, UE9, and T7. Includes a screwdriver. Compatible with LJTick signal-conditioning modules.
Catalog No. NC3357520 Supplier Labjack Corporation Supplier No. CB37
Provides convenient screw terminals for the DB37 connector on the LabJack U6, UE9, and T7. Includes a screwdriver. Compatible with LJTick signal-conditioning modules.