Learn More
American Research Products Inc CB6 Biosimilar
Catalog No. 5000306807
Encompass
CB6 Biosimilar (Anti-SARS-CoV-2) Antibody is a biosimilar antibody with sars-cov-2 reactivity produced in homo sapiens Available in 100 ug size with 7 - 14 business days lead time Ships via gel pack Store at -20 C to -80 C for 12 months in lyophilized form After reconstitution if not intended for use within a month aliquot and store at -80 C (Avoid repeated freezing and thawing) Lyophilized antibodies are shipped at ambient temperature
Catalog No. 50-003-06807 Supplier American Research Products Inc Supplier No. HDBS0011100UG
CB6 Biosimilar (Anti-SARS-CoV-2) Antibody is a biosimilar antibody with sars-cov-2 reactivity produced in homo sapiens Available in 100 ug size with 7 - 14 business days lead time Ships via gel pack Store at -20 C to -80 C for 12 months in lyophilized form After reconstitution if not intended for use within a month aliquot and store at -80 C (Avoid repeated freezing and thawing) Lyophilized antibodies are shipped at ambient temperature