Learn More
PixCell Medical™ HemoScreen CBC Test Kit
Catalog No. 23123804
Change view
Click to view available options
Quantity:
50 Pk.
Catalog No. Quantity
23-123-804 50 Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-123-804 Supplier PixCell Medical™ Supplier No. BCFA00400
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
This item is not returnable. View return policy
HemoScreen CBC Test Kit includes a Cartridge and Sampler to form a self-contained disposable unit. Used for the collection and preparation of blood samples for CBC analysis. The CBC Test Kit is a required consumable for the HemoScreen Analyzer.
  • PixCell's Test Kit design makes blood testing simple using a unique, self-contained, disposable Cartridge and Sampler pairing.
  • The CBC Sampler is a disposable tool used for the collection of venous or capillary blood samples.
  • The CBC Cartridge is a disposable unit that accepts a Sampler-held blood sample and prepares it for HemoScreen CBC analysis.
  • The CBC Cartridge includes all necessary reagents and calibration details for each test.
  • Together, the plastic Cartridge with self-contained reagents and the Sampler holding the correct blood sample volume, make up the CBC Test Kit.
  • The CBC Test Kit is a required consumable for the HemoScreen Analyzer.
  • One Test Kit box includes 50 disposable Test Kits.
Specifications
Type Plastic Cartridge
Quantity 50 Pk.
Dimensions (L x W x H) 3.5 x 2.75 x 0.25 in.