Learn More
Cayman Chemical CBF InhIbItr 10mg
SDP
Catalog No. 502964841
Encompass_Preferred
A small molecule that binds to CBFβ and inhibits its association with Runx1 (IC50 = 3.2 µM); blocks CBFβ-Runx1 interactions in ME-1 cells, acute myeloid leukemia cells harboring dysfunctional CBF, reducing proliferation without toxicity
Catalog No. 50-296-4841 Supplier Cayman Chemical Supplier No. 1677110 MG
A small molecule that binds to CBFβ and inhibits its association with Runx1 (IC50 = 3.2 µM); blocks CBFβ-Runx1 interactions in ME-1 cells, acute myeloid leukemia cells harboring dysfunctional CBF, reducing proliferation without toxicity