Learn More
Cayman Chemical CBFβ Inhibitor, Purity: ≥98%, CAS: 493028-20-9, 25 mg
SDP
Catalog No. 502965473
Encompass_Preferred
A small molecule that binds to CBFβ and inhibits its association with Runx1 (IC50 = 3.2 µM); blocks CBFβ-Runx1 interactions in ME-1 cells, acute myeloid leukemia cells harboring dysfunctional CBF, reducing proliferation without toxicity
Catalog No. 50-296-5473 Supplier Cayman Chemical Supplier No. 1677125 MG
A small molecule that binds to CBFβ and inhibits its association with Runx1 (IC50 = 3.2 µM); blocks CBFβ-Runx1 interactions in ME-1 cells, acute myeloid leukemia cells harboring dysfunctional CBF, reducing proliferation without toxicity